missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
100mL
2.5 L
20 L
500mL
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -58°C |
Color | Colorless |
Density | 0.867g/mL at 25°C |
pH | Neutral |
Boiling Point | 85°C (1013 hPa) |
Flash Point | −2°C (28.4°F) |
Packaging | Glass bottle |
Linear Formula | CH3OCH2CH2OCH3 |
Quantity | 2.5 L |
UN Number | UN2252 |
Show More |
Safety and Handling



Product Identifier
- 1,2-Dimethoxyethane
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,2-dimethoxyethane
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction