Learn More
Mölnlycke™ Biogel™ PI Micro Tech Indicator Underglove
GREENER_CHOICE
Sterile, blue cleanroom undergloves with beaded cuffs and curved fingers are made to be worn in combination with the Biogel™ PI Micro Tech overglove to form a breach indication system. The gloves are suitable for use in aseptic and Class 100 (ISO 5) / EU GMP Grade A environments.
296.64 € - 370.24 €
Specifications
Certifications/Compliance ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102
Certifications/Compliance (Europe) EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420
Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 0.65
View More Specs
Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17885748
GREENER_CHOICE
View Documents
Mölnlycke™
4448955-00
5.5
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17865758
GREENER_CHOICE
View Documents
Mölnlycke™
4448960-00
6
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17855778
GREENER_CHOICE
View Documents
Mölnlycke™
4448965-00
6.5
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17895758
GREENER_CHOICE
View Documents
Mölnlycke™
4448970-00
7
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17875748
GREENER_CHOICE
View Documents
Mölnlycke™
4448975-00
7.5
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17815758
GREENER_CHOICE
View Documents
Mölnlycke™
4448980-00
8
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17825758
GREENER_CHOICE
View Documents
Mölnlycke™
4448985-00
8.5
ONLINE EXCLUSIVE PRICE
530.00 €
370.24 € / 200 pairs
Save 159.76 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
17875778
GREENER_CHOICE
View Documents
Mölnlycke™
4448990-00
9
ONLINE EXCLUSIVE PRICE
424.00 €
296.64 € / 160 pairs
Save 127.36 €
30% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 14-10-2025
Add to basket
Description
Description
  • Blue underglove can be worn together with Biogel™ PI Micro Tech to form a breach indication system
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polyisoprene
  • Exceed industry standard for force at break, tensile strength and elongation
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Specifications
Specifications
ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102
Class 100 (ISO 5)
0.65
Single-use
8.3 mil
Specific
Polyisoprene
0.19 mm
Sterile
0.16 mm
11.8 in.
EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420
No
Blue
Smooth
0.21 mm
Yes
7.5 mil
Powder Free
6.3 mil (Cuff)
24 MPa
30 cm