missing translation for 'onlineSavingsMsg'
Learn More

WHEATON™ Black Screw Cap

Product Code. 30286515 Shop All Wheaton Science Products Products
Click to view available options
Quantity:
140/Pk.
190/Pk.
231/Pk.
264/Pk.
399/Pk.
666/Pk.
882/Pk.
Unit Size:
Pack of 140
Pack of 190
Pack of 231
Pack of 264
Pack of 399
Pack of 666
Pack of 882

Tubular Dram Vial Caps

  • WHEATON™ tubular dram vials are manufactured from clear 5.1 expansion borosilicate glass and use WHEATON™ black polypropylene caps with foil liner
  • Both vials and caps are suitable for storage, display, sample collection and apothecary applications
  • The vials have a buttress thread finish and fit with the buttress thread black polypropylene dram vial caps with foil liner and range from 1.75 mL to 28 mL in volume
  • Dram vial caps are available in black only and are manufactured from polypropylene and have a foil liner, and provide good barrier properties and resistance to hydrocarbons, oils, ketones and alcohol
  • Black polypropylene foil lined dram vial caps are sold separately to match vials sizes and quantity per pack
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Diameter (Metric) Outer 28 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Autoclavable Autoclavable
Quantity 140/Pk.
Type Screw Cap
For Use With (Equipment) Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.