Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Brand:  Fisherbrand™ FB0129110
 View more versions of this product
Product Code. 30928568
  • 93.93 € / Pack of 1000
Description
Rib construction permits stacking without binding
  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.
Specifications
Specifications
50 mL
Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
5 mL
Autoclavable
Yes
Polypropylene
Spout
Beaker
Single-use