Learn More
HTR6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23561-1-AP-20UL
 View more versions of this product
Product Code. 16882385
  • 145.00 € / 20µL
Description
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.
Specifications
Specifications
HTR6
Polyclonal
Unconjugated
HTR6
5 HT 6, 5 HT6, 5 HT6R, 5 hydroxytryptamine receptor 6, HTR6, Serotonin receptor 6
Rabbit
Antigen Affinity Chromatography
RUO
3362
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50406
HTR6
HTR6 Fusion Protein Ag15396
20 μL