missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell HyFlex™ 11-819 Nylon/Spandex ESD Gloves with FORTIX™ Nitrile Foam Coating

Safety gloves with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. Ultra-thin FORTIX™ coating enhances grip, while also maintaining bare hand sensitivity for tasks that require dexterity and precision.
Brand: Ansell 11819070
This item is not returnable.
View return policy
Description
- ESD functionality certified to EN 16350:2014
- Can be used in explosion hazard zones as part of a grounding chain
- Seamless, 18 gauge nylon and spandex liner helps maintain a bare-handed feel for tasks that require dexterity and precision
- FORTIX™ nitrile foam coating enhances grip and provides long lasting protection
- ZONZ™ Comfort Fit Technology ensures gloves are ultra breathable
- Proprietary washing process helps to deliver cleaner, more comfortable gloves
- Touchscreen functionality allows users to work with multiple touchscreen devices without glove removal
- Dermatologically approved by Dermatest™
- Certified by OEKO-TEX™

Specifications
EN 16350:2014, EN 420:2003 + A1:2009 | |
Industrial | |
Blue | |
Specific | |
Nylon, Spandex | |
7 | |
3121A | |
Knit | |
HyFlex™ 11-819 |
Palm | |
Nitrile Foam | |
Reusable | |
Yes | |
ESD Gloves | |
Non-sterile | |
Aerospace, Automotive, Logistics, and Machinery/Equipment | |
Coated | |
18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction