Learn More
MP Biomedicals™ Cellulase from Aspergillus niger
Cellulase from Aspergillus niger catalyzes hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan and cellooligosaccharides cellotriose to cellohexaose
56.60 € - 383.00 €
Specifications
Concentration | >60,000 U/g |
---|---|
Color | Tan |
pH | 4.2 to 5.2 |
For Use With (Application) | Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels |
Content And Storage | 4°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447881
|
MP Biomedicals™
0215058305 |
5 g |
56.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467881
|
MP Biomedicals™
0215058380 |
100 g |
383.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cellulase refers to a group of enzymes which acting together hydrolyze cellulose. The b-glucosidases, including cellobiose are primarily active on the smaller molecular weight cellulose hydrolysates.
- CAS Number: 9012-54-8
- EC Number: 232-734-4
- MDL Number: MFCD00081510
- Format: Tan powder
- Specific activity: >60000U/g
- Readily soluble in water
- Foreign activity: Salmonella Negative
- Optimum pH: 4.2 to 5.2
Cellulase is used in molecular biology, cell biology, textile industry, pulp and paper industry and for commercial food processing in coffee
Specifications
>60,000 U/g | |
4.2 to 5.2 | |
4°C | |
Cellulase |
Tan | |
Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels | |
Aspergillis niger | |
Powder |
Safety and Handling
- cellulase
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : cellulase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.