Learn More
uvex Profastrong NF33 Chemical Protection Gloves
Sensitive NBR chemical protection gloves feature a flocked cotton lining for comfort and provide a high degree of mechanical abrasion resistance.
5.29 € - 12.65 €
Specifications
Color Green
Latex Free Yes
Material NBR
Product Type Chemical-resistant Gloves
EN388 Standard 4101X
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15386922
View Documents Promotion Details
uvex
6012202
8
12.65 €
Each
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
15396922
View Documents Promotion Details
uvex
6012203
9
ONLINE EXCLUSIVE PRICE
12.65 €
5.29 € / Each
Save 7.36 €
58% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
15366922
View Documents Promotion Details
uvex
6012204
10
12.65 €
Each
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
15376922
View Documents Promotion Details
uvex
6012201
7
ONLINE EXCLUSIVE PRICE
12.65 €
5.29 € / Each
Save 7.36 €
58% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Sensitive NBR chemical protection gloves with flocked cotton lining for comfort
  • Embossed structure on the palm enhances grip capabilities in damp and wet areas
  • Nitrile rubber is resistant to oils, grease, acids, and alkalis
  • Gloves provide a tactile feel, secure fit, and mechanical abrasion resistance
  • Ideal for use in various industries including automotive, chemical, and printing
Specifications
Specifications
Green
NBR
4101X
AJKLOT
Gauntlet
33 cm
Yes
Chemical-resistant Gloves
Type A
Bacteria, Fungi WITH Virus Protection
Cotton