missing translation for 'onlineSavingsMsg'
Learn More

uvex ProfiErgo ENB 20 Nitrile-Coated Assembly Gloves

Special NBR coating is resistant to oil and grease, while also being touchscreen compatible. The high-quality cotton interlock liner enhances user comfort, feels pleasant against the skin, and helps to regulate glove temperature.

Brand:  uvex 6014802

11.26 EUR valid until 2025-09-30
Use promo code "27321" to get your promotional price.


 View more versions of this product

Product Code. 15326942

  • ONLINE EXCLUSIVE PRICE
    11.85 €
    7.59 € / Each
    Save 4.26 €
    35% Off
Only null left
In Stock
Edge
Add to basket
This item is not returnable. View return policy

Description

Description

  • Thick, special NBR coating is resistant to oil and grease
  • High-quality cotton interlock liner is comfortable, feels pleasant against the skin, and aids in regulation of the glove climate
  • Enhanced grip capabilities in damp, wet, and oily areas
  • Gloves are suitable for operating touch displays
  • Ideal for a variety of applications including assembly, control work, and maintenance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NBR
Smooth
Yes
Mechanical Protection Gloves
2121X
Interlock Cotton
White, Yellow
Ambidextrous
Cotton
8
Knit
ProfiErgo ENB 20
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.