missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ PUREZERO™ HG3 Light Blue Sterile Nitrile Gloves
BIOPHARMAGREENER_CHOICE

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

Brand:  HALYARD™ CLN923210

385.69 EUR valid until 2025-09-30
Use promo code "24381" to get your promotional price.


 View more versions of this product

Product Code. 17394340

  • 437.00 € / 300 pairs
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.

  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no Phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
  • Backed up by a vertically integrated supply chain
Specifications

Specifications

ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
ASTM D6978
Class 10 (ISO 4)
1
Single-use
4 mil
Specific
Nitrile
0.08 mm
Controlled Environment Gloves
Sterile
0.10 mm
CLN923210
30.5 cm
GMP Grade A/B/C/D
Yes
Yes
Light Blue
Partially Textured
0.10 mm
Yes
3.1 mil
Powder Free
10
4 mil
ISO 9001:2015, EN13485:2016
12 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.