missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HALYARD™ PUREZERO™ HG3 Smooth (SGX) White Nitrile Gloves



High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.
309.00 € - 329.00 €
Specifications
Certifications/Compliance | ISO 3, ISO 4, ISO 5, ISO 6, ISO 7 |
---|---|
Certifications/Compliance (Europe) | GMP Grade A/B/C/D |
Chemotherapeutic Rated | ASTM D6978 |
Chlorinated | Yes |
Cleanroom Class | Class 10 (ISO 4) |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17354330
![]() ![]() |
HALYARD™
CLN3231XS |
X-Small |
309.00 €
Pack of 1000 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
17334330
![]() ![]() |
HALYARD™
CLN3231SM |
Small |
309.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
17324330
![]() ![]() |
HALYARD™
CLN3231MD |
Medium |
309.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
17314330
![]() ![]() |
HALYARD™
CLN3231LG |
Large |
309.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
17344330
![]() ![]() |
HALYARD™
CLN3231XL |
X-Large |
329.00 €
Pack of 1000 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
Description
- Low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination
- Nickel-free
- Powder-free
- Silicone-free
- Latex-free and no phtalates
- High cleanliness
- Low endotoxin levels
- Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
Specifications
ISO 3, ISO 4, ISO 5, ISO 6, ISO 7 | |
ASTM D6978 | |
Class 10 (ISO 4) | |
1 | |
Single-use | |
6 mil | |
Ambidextrous | |
Nitrile | |
0.13 mm | |
Controlled Environment Gloves | |
6 mil | |
ISO 9001:2015, EN13485:2016 | |
30.5 cm |
GMP Grade A/B/C/D | |
Yes | |
Yes | |
White | |
Partially Textured | |
0.16 mm | |
Yes | |
5 mil | |
Powder Free | |
Non-sterile | |
0.16 mm | |
12 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title