missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex Rubiflex S NB60S Chemical Protection Gloves
Long, NBR-coated chemical protection gloves feature reinforced cotton interlock supporting material for applications that require protection against chemicals and mechanical hazards.
41.00 € - 382.00 €
Specifications
Applications | Laboratory |
---|---|
Coating Material | NBR |
Color | Green |
Latex Free | Yes |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17250242
|
uvex
9893055 |
8 |
41.00 €
1 pair |
In Stock
Log in to see stock. ![]() |
|||||
17260242
|
uvex
9893056 |
9 |
382.00 €
10 pairs |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
17280242
|
uvex
9893060 |
10 |
382.00 €
10 pairs |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
17200252
|
uvex
9893199 |
11 |
382.00 €
10 pairs |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
Description
- NBR-coated chemical protection gloves with an extended gauntlet cuff
- Reinforced cotton interlock supporting material provides mechanical protection
- Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
- Ergonomic design is comfortable and highly flexible
- Silicone-free according to imprint test
Specifications
Laboratory | |
Green | |
Nitrile | |
2121X | |
JKOPT | |
Unisex | |
Cotton |
NBR | |
Yes | |
Chemical-resistant Gloves | |
Type B | |
Chemical Handling, Municipal Cleaning, Sandblasting, and Sewer Construction | |
Gauntlet | |
Rubiflex S NB60S |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title