Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Brand:  Memmert™ WTB15B48814
 View more versions of this product
Product Code. 17120315
  • ONLINE EXCLUSIVE PRICE
    129.00 €
    123.00 € / Each
    Save 6.00 €
    4% Off
Only null left
Estimated Shipment: 16-10-2025
Add to basket
Description
Description
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks
Specifications
Specifications
WTB15
Support Basket With Perforated Mounting Shelf (Stainless Steel)
Stainless Steel