Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Brand:  Memmert™ WTB24B48814
 View more versions of this product
Product Code. 17120345
  • ONLINE EXCLUSIVE PRICE
    943.00 DKK
    917.70 DKK / Each
    Save 25.30 DKK
    2% Off
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks
Specifications
Specifications
WTB24
Support Basket With Perforated Mounting Shelf (Stainless Steel)
Stainless Steel