Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Support basket with perforated mounting shelf for conical flasks clips
Brand: Memmert™ WTB35B48815
This item is not returnable.
View return policy
Description
- Use of test tube racks not possible
- Has grid perforations to take clips for conical flasks
Specifications
WTB35 | |
Support Basket With Perforated Mounting Shelf (Stainless Steel) | |
Stainless Steel |