Learn More
Miele Professional™ Water Purification UfZ/1
Product Code. 30533235 Shop All Miele Products
Change view
Click to view available options
Includes:
Installation on Second Cartridge
Unit Size:
1 sheet
Product Code. Includes unitSize
30533235 Installation on Second Cartridge 1 sheet
1 options
Product Code. 30533235 Supplier Miele Professional™ Supplier No. 12684950
For simple and convenient changing of demineralisation cartridges
  • 2 x VA plug connection 3/4, 2 x quick connection 3/4
  • 2 x VA plug connection 3/4 for installation on second cartridge
  • For simple and convenient changing of demineralisation cartridge
TRUSTED_SUSTAINABILITY
Specifications
Product Type Water Connection
For Use With (Equipment) Washer-Disinfectors
Includes Installation on Second Cartridge
Quantity 1 Ea.