missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes
GREENER_CHOICE

Clean surfaces effectively without scratching. Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes are made with fabric knitted from filament polyester for a soft, textured surface.

Brand:  Contec™ AMVS0001

 View more versions of this product

Product Code. 17983752

  • ONLINE EXCLUSIVE PRICE
    481.50 €
    401.25 € / Pack of 1200
    Save 80.25 €
    16% Off
Only null left
Estimated Shipment: 17-12-2025
Edge
Add to basket
This item is not returnable. View return policy

Description

Description

  • Soft, textured wipe surface for effective cleaning without scratching
  • Laser cut, which seals the fibers at edges
  • Made with fabric knitted from filament polyester
  • Packaged in an ISO Class 4 cleanroom to further reduce contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Heatseal Wipe
22.86 cm
22.86 cm
Laser cut
150 Wipes/Bag, 8 Bags/Cs.
9 in.
9 in.
Polyester
9 x 9 in. (22.86 x 22.86 cm)
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.