missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes
GREENER_CHOICE

Clean surfaces effectively without scratching. Contec™ Amplitude™ Standard Weight Heatseal Knit Wipes are made with fabric knitted from filament polyester for a soft, textured surface.

401.25 €

Specifications

Product Type Heatseal Wipe
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
View More Specs

Products 1
Product Code Brand Packaging Quantity Price Quantity & Availability  
Product Code Brand Packaging Quantity Price Quantity & Availability  
17983752
GREENER_CHOICE
View Documents
Contec™
AMVS0001
150 Wipes/Bag, 8 Bags/Cs.
ONLINE EXCLUSIVE PRICE
481.50 €
401.25 € / Pack of 1200
Save 80.25 €
16% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 18-12-2025
Edge
Add to basket
 
Description

Description

  • Soft, textured wipe surface for effective cleaning without scratching
  • Laser cut, which seals the fibers at edges
  • Made with fabric knitted from filament polyester
  • Packaged in an ISO Class 4 cleanroom to further reduce contamination
Specifications

Specifications

Heatseal Wipe
9 x 9 in. (22.86 x 22.86 cm)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.